Learn More
Medchemexpress LLC Phenytoin (5,5-diphenylhydantoin) | 57-41-0 | 99.9% | 1 G
SDP
Catalog No. 5000405344
Encompass_Preferred
Phenytoin (5,5-Diphenylhydantoin) is a potent Voltage-gated Na+ channels (VGSCs) blocker. It has antiepileptic activity and reduces breast tumor growth and metastasis in mice. This product is for research use only and not sold to patients.
  • Potent voltage-gated Na+ channels (VGSCs) blocker
  • Has antiepileptic activity
  • Reduces breast tumor growth and metastasis in mice
Catalog No. 50-004-05344 Supplier Medchemexpress LLC Supplier No. HYB04481G
Phenytoin (5,5-Diphenylhydantoin) is a potent Voltage-gated Na+ channels (VGSCs) blocker. It has antiepileptic activity and reduces breast tumor growth and metastasis in mice. This product is for research use only and not sold to patients.
  • Potent voltage-gated Na+ channels (VGSCs) blocker
  • Has antiepileptic activity
  • Reduces breast tumor growth and metastasis in mice