Learn More
Medchemexpress LLC HY-B0448 500mg , Phenytoin CAS:57-41-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0448500MG
Encompass_Preferred
HY-B0448 500mg Phenytoin CAS: 57-41-0 Phenytoin (5,5-Diphenylhydantoin) is a potent Voltage-gated Na+ channels (VGSCs) blocker. Phenytoin has antiepileptic activity and reduces breast tumour growth and metastasis in mice. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1134