Learn More
Apexbio Technology LLC Phenytoin 57-41-0 5g
SDP
Catalog No. 5000090777
Encompass_Preferred
Phenytoin (57-41-0) is a small-molecule inhibitor targeting voltage-gated sodium channels It is designed to inhibit sodium channel activity thereby regulating neuronal excitability and electrical signaling Phenytoin exerts its biological activity primarily through the stabilization and inhibition of voltage-gated sodium channels by reducing repetitive neuronal firing In in vitro studies Phenytoin demonstrates inhibitory activity with reported IC50 values typically ranging from 40 to 60 M depending upon experimental protocols and neuronal models applied Based on these pharmacological properties Phenytoin holds research potential in neurological studies including epilepsy pathogenesis and sodium channel-targeted therapeutic mechanism evaluation
Catalog No. 50-000-90777 Supplier Apexbio Technology LLC Supplier No. B22715000
Phenytoin (57-41-0) is a small-molecule inhibitor targeting voltage-gated sodium channels It is designed to inhibit sodium channel activity thereby regulating neuronal excitability and electrical signaling Phenytoin exerts its biological activity primarily through the stabilization and inhibition of voltage-gated sodium channels by reducing repetitive neuronal firing In in vitro studies Phenytoin demonstrates inhibitory activity with reported IC50 values typically ranging from 40 to 60 M depending upon experimental protocols and neuronal models applied Based on these pharmacological properties Phenytoin holds research potential in neurological studies including epilepsy pathogenesis and sodium channel-targeted therapeutic mechanism evaluation