Learn More
U.S. Pharmacopeia 2,2-Diphenylglycine, 3060-50-2, MFCD00008048, 50mg
Catalog No. NC1422475
Encompass
Molecular Formula C14H13NO2, Molecular Weight 227.262, Melting Point 473 - 476.6 °F (245 - 247 °C) (decomposes)
Catalog No. NC1422475 Supplier U.S. Pharmacopeia Supplier No. 1535019
Molecular Formula C14H13NO2, Molecular Weight 227.262, Melting Point 473 - 476.6 °F (245 - 247 °C) (decomposes)