Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phenytoin-d10 | 65854-97-9 | MFCD00143404 | 99.5% | 262.33 g/mol | C15H2D10N2O2 | 10 MG
SDP

Catalog No. 5000298287
Encompass_Preferred

Deuterated phenytoin used as a stable-isotope internal standard and analytical reference for quantitative LC- or GC-MS assays. Supplied as a purified solid intended for research use only.

  • Deuterated analog of phenytoin for use as an internal standard in LC- or GC-MS assays.
  • High isotopic labeling to improve accuracy in quantitative analyses.
  • Purity 99.5% as specified by the manufacturer.
  • Molecular weight 262.33 g/mol and CAS number 65854-97-9.
  • Provided as a 10 mg solid vial for laboratory use.
  • For research use only; not for human or clinical applications.

Catalog No. 50-002-98287 Supplier Medchemexpress LLC Supplier No. HYB0448S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Deuterated phenytoin used as a stable-isotope internal standard and analytical reference for quantitative LC- or GC-MS assays. Supplied as a purified solid intended for research use only.

  • Deuterated analog of phenytoin for use as an internal standard in LC- or GC-MS assays.
  • High isotopic labeling to improve accuracy in quantitative analyses.
  • Purity 99.5% as specified by the manufacturer.
  • Molecular weight 262.33 g/mol and CAS number 65854-97-9.
  • Provided as a 10 mg solid vial for laboratory use.
  • For research use only; not for human or clinical applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.