Learn More
Selleck Chemical LLC Phenytoin Sodium S2524-50mg
SDP
Catalog No. 503111636
Encompass_Preferred
Phenytoin Sodium (Aleviatin Diphenylhydantoin sodium Eptoin Phenytoin soluble Prompt Phenytek Dilantin sodium Diphantoine) is an inactive voltage-gated sodium channel stabilizer
Catalog No. 50-311-1636 Supplier Selleck Chemical LLC Supplier No. S252450MG
Phenytoin Sodium (Aleviatin Diphenylhydantoin sodium Eptoin Phenytoin soluble Prompt Phenytek Dilantin sodium Diphantoine) is an inactive voltage-gated sodium channel stabilizer