Learn More
Apexbio Technology LLC Phenytoin sodium 630-93-3 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062400
Encompass_Preferred
Phenytoin sodium is an antiepileptic compound that acts primarily through the stabilization of voltage-gated sodium channels thereby reducing sustained high-frequency neuronal firing Its mechanism involves prolonging the inactive state of these channels limiting repetitive action potentials in neurons Research applications include studying epileptiform activity in vitro and in vivo and examining neuronal network excitability and related neurological pathways Additionally it serves as a standard reference compound in assays evaluating the modulation of sodium channel activities Reported IC50 values for voltage-gated sodium channel inhibition by phenytoin sodium typically range from 10 M to 40 M depending upon experimental conditions and cellular context
Catalog No. 50-000-62400 Supplier Apexbio Technology LLC Supplier No. B22725.1
Phenytoin sodium is an antiepileptic compound that acts primarily through the stabilization of voltage-gated sodium channels thereby reducing sustained high-frequency neuronal firing Its mechanism involves prolonging the inactive state of these channels limiting repetitive action potentials in neurons Research applications include studying epileptiform activity in vitro and in vivo and examining neuronal network excitability and related neurological pathways Additionally it serves as a standard reference compound in assays evaluating the modulation of sodium channel activities Reported IC50 values for voltage-gated sodium channel inhibition by phenytoin sodium typically range from 10 M to 40 M depending upon experimental conditions and cellular context