Learn More
Medchemexpress LLC Phenytoin sodium (5,5-diphenylhydantoin sodium salt) | 630-93-3 | MFCD00069674 | 99.9% | 274.25 | C15H11N2NaO2 | 10 MG
SDP
Catalog No. 5000297291
Encompass_Preferred
Phenytoin sodium is a research-grade sodium salt of 5,5-diphenylhydantoin used as an analytical standard and as a voltage-gated sodium channel inhibitor in biochemical and pharmacological studies. It is supplied in small pack sizes suitable for analytical and in vitro/in vivo formulation work.
Catalog No. 50-002-97291 Supplier Medchemexpress LLC Supplier No. HYB0448AR10MG
Phenytoin sodium is a research-grade sodium salt of 5,5-diphenylhydantoin used as an analytical standard and as a voltage-gated sodium channel inhibitor in biochemical and pharmacological studies. It is supplied in small pack sizes suitable for analytical and in vitro/in vivo formulation work.