Learn More
Apexbio Technology LLC Phenytoin sodium, 5g Cas# 630-93-3
SDP
Supplier:  Apexbio Technology LLC B22725000
Encompass_Preferred
Phenytoin is an inactive voltage-gated sodium channel stabilizer. 630-93-3. MFCD00069674 Phenytoin is an inactive voltage-gated sodium channel stabilizer.Phenytoin is an antiepileptic drug. It is useful to treat partial seizures and generalized tonic-clonic seizures but not primary generalized seizures such as absence seizures or myoclonic se There are other sizes available. Please inqury us for quote.
Catalog No. 50-199-8694