Learn More
Medchemexpress LLC Phenytoin (sodium) | 630-93-3 | 99.9% | 250 MG
SDP
Catalog No. 5000406812
Encompass_Preferred
Phenytoin sodium (Standard) is an analytical standard primarily used for research and analytical applications. It acts as a potent blocker of Voltage-gated Na+ channels (VGSCs) and exhibits antiepileptic activity, with studies showing its potential to reduce breast tumor growth and metastasis. This compound is suitable for qualitative, quantitative, and methodological research experiments, including techniques like HPLC, GC, and MS.
  • Analytical standard for research and analytical applications
  • Potent Voltage-gated Na+ channels (VGSCs) blocker
  • Exhibits antiepileptic activity
  • Reduces breast tumor growth and metastasis in mice (experimental context)
  • Suitable for HPLC, GC, and MS experiments
Catalog No. 50-004-06812 Supplier Medchemexpress LLC Supplier No. 250MGHYB0448AR
Phenytoin sodium (Standard) is an analytical standard primarily used for research and analytical applications. It acts as a potent blocker of Voltage-gated Na+ channels (VGSCs) and exhibits antiepileptic activity, with studies showing its potential to reduce breast tumor growth and metastasis. This compound is suitable for qualitative, quantitative, and methodological research experiments, including techniques like HPLC, GC, and MS.
  • Analytical standard for research and analytical applications
  • Potent Voltage-gated Na+ channels (VGSCs) blocker
  • Exhibits antiepileptic activity
  • Reduces breast tumor growth and metastasis in mice (experimental context)
  • Suitable for HPLC, GC, and MS experiments