Learn More
Medchemexpress LLC Phenytoin (sodium) | 630-93-3 | MFCD00021643 | 99.8% | 5 G
SDP
Catalog No. 5000405447
Encompass_Preferred
Phenytoin sodium is a potent blocker of voltage-gated Na+ channels (VGSCs) with antiepileptic activity. It is an antiepileptic agent useful for treating partial and generalized tonic-clonic seizures, believed to protect against seizures by causing voltage-dependent block of voltage-gated sodium channels. Research indicates it can reduce breast tumor growth and metastasis in mice.
  • Potent blocker of voltage-gated Na+ channels (VGSCs)
  • Exhibits antiepileptic activity
  • Reduces breast tumor growth and metastasis in mice
  • Effective against partial and generalized tonic-clonic seizures
  • Protects against seizures by causing voltage-dependent block of voltage-gated sodium channels
  • Blocks persistent sodium current
  • Class 1b antiarrhythmic
Catalog No. 50-004-05447 Supplier Medchemexpress LLC Supplier No. HYB0448A5G
Phenytoin sodium is a potent blocker of voltage-gated Na+ channels (VGSCs) with antiepileptic activity. It is an antiepileptic agent useful for treating partial and generalized tonic-clonic seizures, believed to protect against seizures by causing voltage-dependent block of voltage-gated sodium channels. Research indicates it can reduce breast tumor growth and metastasis in mice.
  • Potent blocker of voltage-gated Na+ channels (VGSCs)
  • Exhibits antiepileptic activity
  • Reduces breast tumor growth and metastasis in mice
  • Effective against partial and generalized tonic-clonic seizures
  • Protects against seizures by causing voltage-dependent block of voltage-gated sodium channels
  • Blocks persistent sodium current
  • Class 1b antiarrhythmic