Learn More
Medchemexpress LLC Phenytoin sodium | 630-93-3 | MFCD00069674 | 99.8% | 25 MG
SDP
Catalog No. 5000450619
Encompass_Preferred
Phenytoin sodium is a potent blocker of Voltage-gated Na+ channels (VGSCs). It demonstrates antiepileptic activity and has been observed to reduce breast tumor growth and metastasis in mice.
  • Potent Voltage-gated Na+ channels blocker
  • Exhibits antiepileptic activity
  • Reduces breast tumor growth and metastasis in mice
  • Soluble in DMSO for in vitro applications (50 mg/mL)
  • Can be prepared for in vivo use as a clear solution (≥ 2.5 mg/mL)
Catalog No. 50-004-50619 Supplier Medchemexpress LLC Supplier No. 25MGHYB0448AR
Phenytoin sodium is a potent blocker of Voltage-gated Na+ channels (VGSCs). It demonstrates antiepileptic activity and has been observed to reduce breast tumor growth and metastasis in mice.
  • Potent Voltage-gated Na+ channels blocker
  • Exhibits antiepileptic activity
  • Reduces breast tumor growth and metastasis in mice
  • Soluble in DMSO for in vitro applications (50 mg/mL)
  • Can be prepared for in vivo use as a clear solution (≥ 2.5 mg/mL)