Learn More
Medchemexpress LLC Phenytoin sodium (Standard) | 630-93-3 | 99.9% | 100 MG
SDP
Catalog No. 5000405342
Encompass_Preferred
Phenytoin sodium (Standard) is the analytical standard of Phenytoin (sodium), intended for research and analytical applications. It acts as a potent Voltage-gated Na+ channels (VGSCs) blocker and has antiepileptic activity, demonstrating potential to reduce breast tumor growth and metastasis in mice.
  • Analytical standard for research.
  • Potent blocker of voltage-gated Na+ channels (VGSCs).
  • Exhibits antiepileptic activity.
  • Reduces breast tumor growth and metastasis in mice.
  • Reference standard for assay.
  • Suitable for HPLC, GC, and MS experiments.
Catalog No. 50-004-05342 Supplier Medchemexpress LLC Supplier No. HYB0448AR100MG
Phenytoin sodium (Standard) is the analytical standard of Phenytoin (sodium), intended for research and analytical applications. It acts as a potent Voltage-gated Na+ channels (VGSCs) blocker and has antiepileptic activity, demonstrating potential to reduce breast tumor growth and metastasis in mice.
  • Analytical standard for research.
  • Potent blocker of voltage-gated Na+ channels (VGSCs).
  • Exhibits antiepileptic activity.
  • Reduces breast tumor growth and metastasis in mice.
  • Reference standard for assay.
  • Suitable for HPLC, GC, and MS experiments.