Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenytoin Sodium, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18605148 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
12 kg
Packaging:
Amber Glass Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-148 25 g Amber Glass Bottle
18-605-149 100 g Amber Glass Bottle
18-605-150 500 g Amber Glass Bottle
18-605-151 2.5 kg Poly Pail
18-605-152 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-605-148 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P109425GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

P1094, 630-93-3, C15H11N2NaO2

Spectrum™ Chemical Phenytoin Sodium, USP is an anticonvulsant or antiepileptic drug used to prevent and control seizures. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 630-93-3
Molecular Formula C15H11N2NaO2
Molecular Weight (g/mol) 274.26
MDL Number MFCD00069674,MFCD00005264
IUPAC Name sodium 2,5-dioxo-4,4-diphenylimidazolidin-1-ide

Specifications

CAS Max % 1
Quantity 25 g
Grade USP
Loss on Drying 0.025
Packaging Amber Glass Bottle
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.