Learn More
Description
Specifications
Specifications
| CAS Max % | 1 |
| Physical Form | Crystalline Powder or Needles |
| Quantity | 12 kg |
| Grade | USP |
| Loss on Drying | 0.025 |
| Packaging | Amber Glass Bottle |
| Chemical Name or Material | Phenytoin Sodium |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
