Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medisca Phenytoin Sodium USP/EP
SDP

Catalog No. 14185426 Shop All Medisca Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-185-426 500 g
14-185-106 100 g
14-185-383 1 kg
14-185-422 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 14-185-426 Supplier Medisca Supplier No. 021608
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phenytoin Sodium USP / EP is a white or almost white, very hygroscopic, crystalline powder. It is odorless.

Phenytoin Sodium USP / EP is a white or almost white, very hygroscopic, crystalline powder. It is odorless.

Specifications

Chemical Name or Material Phenytoin Sodium USP/EP
Packaging Jar, 1500mL, White, HDPE, Round, 89-400, TE Pano Neck
CAS 630-93-3
Type Drug Standard
Molecular Weight (g/mol) 274.2539
Quantity 500 g
Percent Purity 98 to 102 %
Grade EP, USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.