Learn More
Cayman Chemical PheophorbIde a 100mg
SDP
Catalog No. 502972172
Encompass_Preferred
A product of chlorophyll breakdown that has been used as a photosensitizer in photodynamic therapy for the treatment of cancer; inhibits glioblastoma cell growth (IC50 = 2.8 UG/ml) and displays antiproliferative activity against melanoma, breast, and lung cancer cells in vitro at 100 UG/ml
Catalog No. 50-297-2172 Supplier Cayman Chemical Supplier No. 16072100 MG
A product of chlorophyll breakdown that has been used as a photosensitizer in photodynamic therapy for the treatment of cancer; inhibits glioblastoma cell growth (IC50 = 2.8 UG/ml) and displays antiproliferative activity against melanoma, breast, and lung cancer cells in vitro at 100 UG/ml