Learn More
Cayman Chemical Phepropeptin D, Purity: ≥98%, CAS: 396729-26-3, 1 mg
SDP
Catalog No. 502887142
Encompass_Preferred
A cyclic peptide with proteasome inhibitory activity; inhibits isolated mouse liver proteasomes (IC50 = 7.8 µg/ml)
Catalog No. 50-288-7142 Supplier Cayman Chemical Supplier No. 100081111 MG
A cyclic peptide with proteasome inhibitory activity; inhibits isolated mouse liver proteasomes (IC50 = 7.8 µg/ml)