Learn More
Aviva Systems Biology Corp PHF11 Antibody - N-terminal region (ARP39228_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39228_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PHF11. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the n terminal region of human PHF11. Alternate names: APY, BCAP, IGEL, IGER, IGHER, NY-REN-34, NYREN34, RP11-185C18.3ApplicationWB. Species reactivity: Cow, Horse, Human
Catalog No. 50-014-56950