Learn More
Aviva Systems Biology Corp PHF12 Antibody - C-terminal region (ARP50416_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50416_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PHF12. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human PHF12. Alternate names: FLJ34122, KIAA1523, MGC131914, PF1ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60338