Learn More
Aviva Systems Biology Corp PHF13 Antibody - middle region (ARP51228_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51228_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PHF13. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human PHF13. Alternate names: MGC43399, PHF5, SPOC1ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-565