Learn More
Aviva Systems Biology Corp JADE1 Antibody - C-terminal region (ARP34715_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34715_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PHF17. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human PHF17. Alternate names: FLJ22479, JADE1, KIAA1807ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55529