Learn More
Aviva Systems Biology Corp PHF2 Antibody - middle region (ARP33030_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33030_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PHF2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PHF2. Alternate names: GRC5, JHDM1E, KIAA0662, MGC176680, CENP-35ApplicationWB. Species reactivity: Cow, Horse, Human, Rabbit, Yeast
Catalog No. 50-014-54856