Learn More
Aviva Systems Biology Corp PHF20 Antibody - C-terminal region (ARP35917_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35917_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PHF20. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human PHF20. Alternate names: RP5-1121G12.1, C20orf104, FLJ33479, GLEA2, HCA58, NZF, TZPApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Pig, Rabbit
Catalog No. 50-014-55924