Learn More
Aviva Systems Biology Corp Phf23 Antibody - N-terminal region (ARP51233_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51233_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Phf23. It was validated on Western Blot. . Alternate names: RP23-172M21.13-002, 2410141M05Rik, JUNE-1, RP23-172M21.13ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-566