Learn More
Aviva Systems Biology Corp PHF6 Antibody - C-terminal region (ARP51236_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51236_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PHF6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human PHF6. Alternate names: BORJ, MGC14797, BFLS, CENP-31ApplicationWB. Species reactivity: Human
Catalog No. 50-014-60523