Learn More
Medchemexpress LLC PHF6 TFA | 99.5% | 748.91 | 5 MG
SDP
Catalog No. 5000399174
Encompass_Preferred
PHF6 (VQIVYK) TFA is a self-assembly sequence capable of initiating the full-length tau protein aggregation. PHF6 TFA is mapped to the third microtubule-binding repeat region of the tau protein.
  • Aggregated tau-PHF6 (VQIVYK) TFA potentiates NLRP3 inflammasome expression and autophagy in human microglial cells.
Catalog No. 50-003-99174 Supplier Medchemexpress LLC Supplier No. HYP4808A5MG
PHF6 (VQIVYK) TFA is a self-assembly sequence capable of initiating the full-length tau protein aggregation. PHF6 TFA is mapped to the third microtubule-binding repeat region of the tau protein.
  • Aggregated tau-PHF6 (VQIVYK) TFA potentiates NLRP3 inflammasome expression and autophagy in human microglial cells.