Learn More
Hach Company Radiometer Analytical PHG301-9 Glass pH Electrode (screw cap)
Catalog No. 501997570 Shop All Hach Company Products
Encompass
The Radiometer PHG301 pH Electrode has a glass body and should be used with Ag/AgCl or calomel reference electrodes for general purpose applications. The PHG301 is a screw cap electrode and requires the CL114 cable (part no. A94L114) for compatibilty with Hach Sension+ laboratory pH meters (or any meter with a BNC connection).
Catalog No. 50-199-7570 Supplier Hach Company Supplier No. E11M001
The Radiometer PHG301 pH Electrode has a glass body and should be used with Ag/AgCl or calomel reference electrodes for general purpose applications. The PHG301 is a screw cap electrode and requires the CL114 cable (part no. A94L114) for compatibilty with Hach Sension+ laboratory pH meters (or any meter with a BNC connection).