Learn More
Hach Company Radiometer Analytical PHG311-9 Glass pH Electrode for Alkaline Samples (alkaline glass, screw cap)
Catalog No. 501997571 Shop All Hach Company Products
Encompass
The Radiometer PHG311 pH Electrode has a glass body and pH membrane specifc for alkaline samples. The PHG311 should be used with Ag/AgCl or calomel reference electrodes for general purpose applications. The PHC311-9 is a screw cap electrode and requires the CL114 cable (part no. A94L114) for compatibilty with Hach Sension+ laboratory pH meters (or any meter with a BNC connection).
Catalog No. 50-199-7571 Supplier Hach Company Supplier No. E11M004
The Radiometer PHG311 pH Electrode has a glass body and pH membrane specifc for alkaline samples. The PHG311 should be used with Ag/AgCl or calomel reference electrodes for general purpose applications. The PHC311-9 is a screw cap electrode and requires the CL114 cable (part no. A94L114) for compatibilty with Hach Sension+ laboratory pH meters (or any meter with a BNC connection).