Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phgdh-inactive | 1914971-16-6 | 99.9% | 1 MG
SDP

Catalog No. 5000232464
Encompass_Preferred

PHGDH-inactive is a research compound supplied as an inactive negative control for PHGDH inhibitor studies, intended for research use only. It is available as small solid quantities and as a DMSO solution, with manufacturer-provided solubility and storage recommendations.

  • Has no activity against phosphoglycerate dehydrogenase (PHGDH).
  • High purity (99.9%).
  • Molecular weight 327.45 g·mol⁻¹.
  • Soluble in DMSO; manufacturer provides in vivo formulation protocols.
  • Available as small solid quantities and as a 10 mM DMSO solution.

Catalog No. 50-002-32464 Supplier Medchemexpress LLC Supplier No. HY1217331MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PHGDH-inactive is a research compound supplied as an inactive negative control for PHGDH inhibitor studies, intended for research use only. It is available as small solid quantities and as a DMSO solution, with manufacturer-provided solubility and storage recommendations.

  • Has no activity against phosphoglycerate dehydrogenase (PHGDH).
  • High purity (99.9%).
  • Molecular weight 327.45 g·mol⁻¹.
  • Soluble in DMSO; manufacturer provides in vivo formulation protocols.
  • Available as small solid quantities and as a 10 mM DMSO solution.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.