Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PHGDH-inactive | 1914971-16-6 | 99.9% | 1 ML
SDP

Catalog No. 5000230229
Encompass_Preferred

PHGDH-inactive is a chemically defined negative control compound used to validate PHGDH inhibitor assays. It is supplied in both ready-to-use solution and solid formats for in vitro research use only.

  • Used as a negative control in PHGDH inhibitor assays.
  • Available as a 10 mM solution in DMSO and as solid powders.
  • High chemical purity (about 99.9%).
  • Soluble in DMSO at approximately 83.33 mg/mL; ultrasonic assistance may be required.
  • Store at 4°C protected from light; in solution, store long-term at -80°C.
  • For research use only; not for human or clinical use.

Catalog No. 50-002-30229 Supplier Medchemexpress LLC Supplier No. HY12173310MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PHGDH-inactive is a chemically defined negative control compound used to validate PHGDH inhibitor assays. It is supplied in both ready-to-use solution and solid formats for in vitro research use only.

  • Used as a negative control in PHGDH inhibitor assays.
  • Available as a 10 mM solution in DMSO and as solid powders.
  • High chemical purity (about 99.9%).
  • Soluble in DMSO at approximately 83.33 mg/mL; ultrasonic assistance may be required.
  • Store at 4°C protected from light; in solution, store long-term at -80°C.
  • For research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.