Learn More
Cayman Chemical PHGDH-inactive, Purity: ≥98%, CAS: 1914971-16-6, 1 mg
SDP
Catalog No. 502984299
Encompass_Preferred
An inactive analog of the PHGDH inhibitors NCT-502 and NCT-503
Catalog No. 50-298-4299 Supplier Cayman Chemical Supplier No. 197171 MG
An inactive analog of the PHGDH inhibitors NCT-502 and NCT-503