Learn More
Cayman Chemical PHGDH-inactive, Purity: ≥98%, CAS: 1914971-16-6, 25 mg
SDP
Catalog No. 502985418
Encompass_Preferred
An inactive analog of the PHGDH inhibitors NCT-502 and NCT-503
Catalog No. 50-298-5418 Supplier Cayman Chemical Supplier No. 1971725 MG
An inactive analog of the PHGDH inhibitors NCT-502 and NCT-503