Learn More
Selleck Chemical LLC PHI-101-E1437-25MG
SDP
Catalog No. 503137118
Encompass_Preferred
PHI-101 is an orally active potent third-generation inhibitor of FLT3 that overcomes resistance to multiple drug-resistant mutations It has potential for research in relapsed or refractory acute myeloid leukemia (AML)
Catalog No. 50-313-7118 Supplier Selleck Chemical LLC Supplier No. E143725MG
PHI-101 is an orally active potent third-generation inhibitor of FLT3 that overcomes resistance to multiple drug-resistant mutations It has potential for research in relapsed or refractory acute myeloid leukemia (AML)