Learn More
New England Biolabs, Inc. phi29 DNA Polymerase – 250 units
SDP
Catalog No. 50811699
Encompass
phi29 DNA Polymerase is the replicative polymerase from the Bacillus subtilis phage phi29. This polymerase has exceptional strand displacement and processive synthesis properties. The polymerase has an inherent 3' to 5' proofreading exonuclease activity.
Catalog No. 50-811-699 Supplier New England Biolabs, Inc. Supplier No. M0269S
phi29 DNA Polymerase is the replicative polymerase from the Bacillus subtilis phage phi29. This polymerase has exceptional strand displacement and processive synthesis properties. The polymerase has an inherent 3' to 5' proofreading exonuclease activity.