Learn More
Qiagen Beverly Inc PHI29 DNA POLYMERASE DNAHIGH
Catalog No. 50305920
Encompass
DNA Polymerase, Phi-29 DNA Polymerase High Conc.; Conc. 100,000 U/ul; 2,000 U; incl. 10X buffer
Catalog No. 50-305-920 Supplier Qiagen Beverly Inc Supplier No. P7020HCL
DNA Polymerase, Phi-29 DNA Polymerase High Conc.; Conc. 100,000 U/ul; 2,000 U; incl. 10X buffer