Learn More
System Biosciences LLC PHIC31 INTEGRASE EXP. PLASMIDPhiC31 Integrase Expression Plasmid = 10ug
SDP
Catalog No. NC2038597
Encompass_Preferred
PinPoint genome editing projects with this vector that expresses PhiC31 Integrase for one-step single-copy transgenesis. Designed to transiently deliver PhiC31 Integrase for your PhiC31 and PinPoint genome engineering studies, the PhiC31 Integrase Expression Plasmid is an essential component of these one- and two-step non-viral gene delivery systems. PhiC31 integrase can integrate a plasmid of any size, as a single copy, and requires no cofactors. The integrated transgenes are stably expressed and heritable.
Catalog No. NC2038597 Supplier System Biosciences LLC Supplier No. FC200PA1
PinPoint genome editing projects with this vector that expresses PhiC31 Integrase for one-step single-copy transgenesis. Designed to transiently deliver PhiC31 Integrase for your PhiC31 and PinPoint genome engineering studies, the PhiC31 Integrase Expression Plasmid is an essential component of these one- and two-step non-viral gene delivery systems. PhiC31 integrase can integrate a plasmid of any size, as a single copy, and requires no cofactors. The integrated transgenes are stably expressed and heritable.