Learn More
Medchemexpress LLC HY-108637 5mg Medchemexpress, PhiKan 083 CAS:880813-36-5 Purity:>98%
SDP
Catalog No. 501932326
Encompass_Preferred
Medchemexpress, HY-108637 5mg PhiKan 083 CAS:880813-36-5 PhiKan 083 is a carbazole derivative, which binds to the surface cavity and stabilizes Y220C (a p53 mutant), with a K d of 167 μM [1] , and a relative binding affinity ( K d ) of 150 μM in Ln229 cells [3] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2326 Supplier Medchemexpress LLC Supplier No. HY1086375MG
Medchemexpress, HY-108637 5mg PhiKan 083 CAS:880813-36-5 PhiKan 083 is a carbazole derivative, which binds to the surface cavity and stabilizes Y220C (a p53 mutant), with a K d of 167 μM [1] , and a relative binding affinity ( K d ) of 150 μM in Ln229 cells [3] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.