Learn More
Medchemexpress LLC HY-108637A 5mg Medchemexpress, PhiKan 083 hydrochloride CAS:1050480-30-2 Purity:>98%
SDP
Catalog No. 501968600
Encompass_Preferred
Medchemexpress, HY-108637A 5mg PhiKan 083 HCl CAS:1050480-30-2 PhiKan 083 hydrochloride is a carbazole derivative, which binds to the surface cavity and stabilizes Y220C (a p53 mutant), with a Kd of 167 μM, and a relative binding affinity (Kd) of 150 μM in Ln229 cells[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8600 Supplier Medchemexpress LLC Supplier No. HY108637A5MG
Medchemexpress, HY-108637A 5mg PhiKan 083 HCl CAS:1050480-30-2 PhiKan 083 hydrochloride is a carbazole derivative, which binds to the surface cavity and stabilizes Y220C (a p53 mutant), with a Kd of 167 μM, and a relative binding affinity (Kd) of 150 μM in Ln229 cells[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.