Learn More
Medchemexpress LLC PhiKan 083 hydrochlo | 1050480-30-2 | 99.3% | 274.79 | 10 MG
SDP
Catalog No. 5000346311
Encompass_Preferred
PhiKan 083 hydrochloride is a carbazole derivative that functions as a p53 Y220C stabilizer. It binds to the surface cavity of the Y220C (a p53 mutant) with a Kd of 167 μM and demonstrates a relative binding affinity (Kd) of 150 μM in Ln229 cells. This compound has been shown to slow down the thermal denaturation rate of p53 Y220C.
  • Binds to the surface cavity of p53 Y220C mutant.
  • Stabilizes p53 Y220C.
  • Exhibits a Kd of 167 μM for p53 Y220C.
  • Shows a relative binding affinity of 150 μM in Ln229 cells.
  • Slows the thermal denaturation rate.
  • Reduces cell viability of engineered Ln229 cell variants.
  • Enhances pro-apoptotic activity in Ln229 cells when combined with NSC 123127.
Catalog No. 50-003-46311 Supplier Medchemexpress LLC Supplier No. HY108637A50MG
PhiKan 083 hydrochloride is a carbazole derivative that functions as a p53 Y220C stabilizer. It binds to the surface cavity of the Y220C (a p53 mutant) with a Kd of 167 μM and demonstrates a relative binding affinity (Kd) of 150 μM in Ln229 cells. This compound has been shown to slow down the thermal denaturation rate of p53 Y220C.
  • Binds to the surface cavity of p53 Y220C mutant.
  • Stabilizes p53 Y220C.
  • Exhibits a Kd of 167 μM for p53 Y220C.
  • Shows a relative binding affinity of 150 μM in Ln229 cells.
  • Slows the thermal denaturation rate.
  • Reduces cell viability of engineered Ln229 cell variants.
  • Enhances pro-apoptotic activity in Ln229 cells when combined with NSC 123127.