Learn More
Medchemexpress LLC HY-104020A 25mg Medchemexpress, Philanthotoxin 74 dihydrochloride PhTx 74 dihydrochloride CAS:1227301-51-0 Purity:98%
SDP
Catalog No. 501637225
Encompass_Preferred
Medchemexpress,HY-104020A 25mg Philanthotoxin 74 dihydrochloride PhTx 74 dihydrochloride CAS:1227301-51-0 Formula:C24H44Cl2N4O3 Philanthotoxin 74 dihydrochloride (PhTx 74) is an AMPAR antagonist; inhibits GluR3 and GluR1 with IC50s of 263 and 296 nM, respectively. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-7225 Supplier Medchemexpress LLC Supplier No. HY104020A25MG
Medchemexpress,HY-104020A 25mg Philanthotoxin 74 dihydrochloride PhTx 74 dihydrochloride CAS:1227301-51-0 Formula:C24H44Cl2N4O3 Philanthotoxin 74 dihydrochloride (PhTx 74) is an AMPAR antagonist; inhibits GluR3 and GluR1 with IC50s of 263 and 296 nM, respectively. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.