Learn More
Medchemexpress LLC HY-104020A 1mg Medchemexpress, Philanthotoxin 74 dihydrochloride CAS:1227301-51-0 Purity:>98%
SDP
Catalog No. 501968189
Encompass_Preferred
Medchemexpress, HY-104020A 1mg Philanthotoxin 74 diHCl CAS:1227301-51-0 Philanthotoxin 74 dihydrochloride (PhTx 74) is an AMPAR antagonist; inhibits GluR3 and GluR1 with IC50s of 263 and 296 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8189 Supplier Medchemexpress LLC Supplier No. HY104020A1MG
Medchemexpress, HY-104020A 1mg Philanthotoxin 74 diHCl CAS:1227301-51-0 Philanthotoxin 74 dihydrochloride (PhTx 74) is an AMPAR antagonist; inhibits GluR3 and GluR1 with IC50s of 263 and 296 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.