Learn More
Medchemexpress LLC Philanthotoxin 74 dihydrochloride | 1227301-51-0 | 99.7% | 507.54 g/mol | C24H44Cl2N4O3 | 1 ML
SDP
Catalog No. 5000242629
Encompass_Preferred
Philanthotoxin 74 dihydrochloride is a synthetic polyamine-derived AMPA receptor antagonist used in pharmacology and electrophysiology research. It selectively inhibits AMPAR subunits with reported IC50 values of 263 nM (GluR3) and 296 nM (GluR1), and is supplied as a high-purity reagent for in vitro assays.
  • High purity suitable for research applications.
  • Supplied as a 10 mM solution in DMSO (1 ML) for ready use in assays.
  • Selective AMPAR antagonist useful for GluR1 and GluR3 studies.
  • Appropriate for electrophysiology and pharmacology experiments.
  • Includes chemical formula and molecular weight for dose calculations.
  • For research use only; not for human or diagnostic use.
Catalog No. 50-002-42629 Supplier Medchemexpress LLC Supplier No. HY104020A10MM 1ML
Add to Cart
Add to Cart
Philanthotoxin 74 dihydrochloride is a synthetic polyamine-derived AMPA receptor antagonist used in pharmacology and electrophysiology research. It selectively inhibits AMPAR subunits with reported IC50 values of 263 nM (GluR3) and 296 nM (GluR1), and is supplied as a high-purity reagent for in vitro assays.
  • High purity suitable for research applications.
  • Supplied as a 10 mM solution in DMSO (1 ML) for ready use in assays.
  • Selective AMPAR antagonist useful for GluR1 and GluR3 studies.
  • Appropriate for electrophysiology and pharmacology experiments.
  • Includes chemical formula and molecular weight for dose calculations.
  • For research use only; not for human or diagnostic use.