Learn More
Electron Microscopy Sciences Philips SEM 4-Position Multi-Sample Holder
SDP
Catalog No. 501930541
Encompass_Preferred
A single tilt multi-grid holder. Will accommodate 4 grids with easy reproducible movement between each specimen. It is ideal for scanning through serial samples or for comparing against standards. Grids are held in place by easy to remove circlips. Comes complete with loading stand, circlip extraction tool, and is supplied in a mahogany box. Suitable for 400 and non-compustage microscopes.
Catalog No. 50-193-0541 Supplier Electron Microscopy Sciences Supplier No. EMS020
Add to Cart
Add to Cart
Request a Quote
A single tilt multi-grid holder. Will accommodate 4 grids with easy reproducible movement between each specimen. It is ideal for scanning through serial samples or for comparing against standards. Grids are held in place by easy to remove circlips. Comes complete with loading stand, circlip extraction tool, and is supplied in a mahogany box. Suitable for 400 and non-compustage microscopes.