Learn More
Electron Microscopy Sciences Philips Bulk Sample Holder for Compustage
SDP
Catalog No. 501930545
Encompass_Preferred
A single-tilt multi-functional sample holder. For use in either SEM or TEM modes. Includes slit serial section tip, 10 bulk, and a TEM 3.05 mm grid holder for use with TEM or STEM. Comes supplied in its own mahogany box, with a loading stand, and a circlip extraction tool.
Catalog No. 50-193-0545 Supplier Electron Microscopy Sciences Supplier No. EMS054
Add to Cart
Add to Cart
Request a Quote
A single-tilt multi-functional sample holder. For use in either SEM or TEM modes. Includes slit serial section tip, 10 bulk, and a TEM 3.05 mm grid holder for use with TEM or STEM. Comes supplied in its own mahogany box, with a loading stand, and a circlip extraction tool.