Learn More
Electron Microscopy Sciences Philips Single Tilt Sample Holder
SDP
Catalog No. 501930542
Encompass_Preferred
A single tilt single sample holder. Will accommodate 3 mm grids. Grids are held in place by easy to remove circlips. Comes complete with loading stand, circlip extraction tool, and is supplied in its own box. Suitable for 400 and non-compustage microscopes.
Catalog No. 50-193-0542 Supplier Electron Microscopy Sciences Supplier No. EMS021
Add to Cart
Add to Cart
Request a Quote
A single tilt single sample holder. Will accommodate 3 mm grids. Grids are held in place by easy to remove circlips. Comes complete with loading stand, circlip extraction tool, and is supplied in its own box. Suitable for 400 and non-compustage microscopes.