Learn More
Bulbs Com PHILLIPS 4W 6IN T5 TUV BULB
SDP
Catalog No. NC1478038
Encompass
Philips 4W 6in T5 TUV Germicidal Fluorescent TubeTUV4T5 (G4T5)Produces UVC radiation to kill microscopic organisms. CAUTION: can be very harmful to eyes and skin. DO NOT expose eyes or unprotected skin to these lamps. SERIOUS injury and blindness could result.
Catalog No. NC1478038 Supplier Bulbs Com Supplier No. 363713
Philips 4W 6in T5 TUV Germicidal Fluorescent TubeTUV4T5 (G4T5)Produces UVC radiation to kill microscopic organisms. CAUTION: can be very harmful to eyes and skin. DO NOT expose eyes or unprotected skin to these lamps. SERIOUS injury and blindness could result.