Learn More
Grainger Moody Tool Fixed ESD-Safe Short Phillips Driver, 0.080 in., 1 ea.
Catalog No. 502774346
Encompass_Preferred
This Phillips screwdriver is a fixed, ESD-safe tool designed for precision tasks. It features a short handle and an .080 in. tip, making it ideal for delicate applications where electrostatic discharge protection is required.
  • Fixed, ESD-safe short handle design
  • Phillips tip size .080 in.
  • TAA compliant
  • ANSI certified
  • Packaged in a hanging poly bag
Catalog No. 50-277-4346 Supplier Grainger Supplier No. 171Y05
This Phillips screwdriver is a fixed, ESD-safe tool designed for precision tasks. It features a short handle and an .080 in. tip, making it ideal for delicate applications where electrostatic discharge protection is required.
  • Fixed, ESD-safe short handle design
  • Phillips tip size .080 in.
  • TAA compliant
  • ANSI certified
  • Packaged in a hanging poly bag