Learn More
Electron Microscopy Sciences Phillips Screwdriver Phillips Screwdriver Set
SDP
Catalog No. 5027843
Encompass_Preferred
A very handy set of Phillips screwdrivers which feature one of the smallest sizes available (1.20 mm). A knurled body with hexagonal heads and tempered steel blades. All are 4 1/2" length. Sizes: 1.20mm, 1.50mm, 2.00mm, and 2.50mm.
Catalog No. 50-278-43 Supplier Electron Microscopy Sciences Supplier No. 6207700
Add to Cart
Add to Cart
A very handy set of Phillips screwdrivers which feature one of the smallest sizes available (1.20 mm). A knurled body with hexagonal heads and tempered steel blades. All are 4 1/2" length. Sizes: 1.20mm, 1.50mm, 2.00mm, and 2.50mm.