Learn More
Electron Microscopy Sciences Phillips Screwdriver Size 1.2 mm
SDP
Supplier:  Electron Microscopy Sciences 6207712
Encompass_Preferred
A very handy set of Phillips screwdrivers which feature one of the smallest sizes available (1.20 mm). A knurled body with hexagonal heads and tempered steel blades. All are 4 1/2" length.
Catalog No. 50-278-44